Learn More
2-Acetamido-3-O-(D-1-carboxyethyl)-2-deoxy-2-D-glucose, TRC
Product Code. 30179069
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30179069 10 mg 10mg
30178957 25 mg 25mg
30179192 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179069 Supplier Toronto Research Chemicals Supplier No. TRCA14826510MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 10597-89-4
Molecular Formula C11H19NO8
Molecular Weight (g/mol) 293.27
Synonym N-Acetylmuramic Acid;
IUPAC Name (2R)-2-[(2R,3R,4R,5R)-2-acetamido-4,5,6-trihydroxy-1-oxohexan-3-yl]oxypropanoic acid
SMILES C[C@@H](O[C@@H]([C@H](O)[C@H](O)CO)[C@@H](NC(=O)C)C=O)C(=O)O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C11H19NO8/c1-5(11(18)19)20-10(9(17)8(16)4-14)7(3-13)12-6(2)15/h3,5,7-10,14,16-17H,4H2,1-2H3,(H,12,15)(H,18,19)/t5-,7+,8-,9-,10-/m1/s1
Formula Weight 293.1111
Chemical Name or Material 2-Acetamido-3-O-(D-1-carboxyethyl)-2-deoxy-2-D-glucose