Learn More
2-Acetamido-2-deoxy-D-mannono-1,4-lactone, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Acetamido-2-deoxy-D-mannono-1,4-lactone |
| CAS | 28876-37-1 |
| Molecular Formula | C8 H13 N O6 |
| Quantity | 25 mg |
| Synonym | D-Mannonic acid, 2-(acetylamino)-2-deoxy-, γ-lactone, Mannonic acid, 2-acetamido-2-deoxy-, γ-lactone, D- (8CI), 2-Acetamido-2-deoxy-D-mannono-1,4-lactone, NSC 297283 |
| InChI Formula | InChI=1S/C8H13NO6/c1-3(11)9-5-6(13)7(4(12)2-10)15-8(5)14/h4-7,10,12-13H,2H2,1H3,(H,9,11)/t4-,5+,6-,7-/m1/s1 |
| SMILES | CC(=O)N[C@H]1[C@@H](O)[C@H](OC1=O)[C@H](O)CO |
| IUPAC Name | N-[(3S,4R,5S)-5-[(1R)-1,2-dihydroxyethyl]-4-hydroxy-2-oxooxolan-3-yl]acetamide |
| Molecular Weight (g/mol) | 219.19 |
| Formula Weight | 219.0743 |