Learn More
MP Biomedicals™ 2-8 CELLsium™ Medium for Short-Term Cell Storage
Product Code. 16309865 Shop All MP Biomedical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. 16309865
Brand: MP Biomedicals™ 092780354
2-8 CELLsius is a cytoprotective, protein free, ready to use bio-solution without DMSO for the short-term storage of cells or tissue.
  • For short-term storage, add cold (2 to 8 °C) 2-8 CELLsium at a recommended cell concentration of 1 x 106 mL to 8 x 106 mL
  • The 2-8 CELLsium can be removed after centrifugation, cell concentration adjusted and transferred to an appropriate culture flask with growth medium
  • It will keep the cells healthy while in a temporary senescent phase, unlike regular growth media, which cannot
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Sterility Sterile
For Use With (Application) Short-term cell storage
Quantity 500 mL
Content And Storage Stored at 2 to 8°C