Learn More
2,7-Dinitrofluorene, TRC
Specifications
Specifications
| Chemical Name or Material | 2,7-Dinitrofluorene |
| CAS | 5405-53-8 |
| Molecular Formula | C13 H8 N2 O4 |
| Quantity | 10 g |
| Synonym | 9H-Fluorene, 2,7-dinitro- (9CI, ACI), 2,7-Dinitro-9H-fluorene (ACI), Fluorene, 2,7-dinitro- (7CI, 8CI), 2,7-Dinitrofluorene, NSC 5180 |
| InChI Formula | InChI=1S/C13H8N2O4/c16-14(17)10-1-3-12-8(6-10)5-9-7-11(15(18)19)2-4-13(9)12/h1-4,6-7H,5H2 |
| SMILES | [O-][N+](=O)c1ccc2c(Cc3cc(ccc23)[N+](=O)[O-])c1 |
| IUPAC Name | 2,7-dinitro-9H-fluorene |
| Molecular Weight (g/mol) | 256.21 |
| Formula Weight | 256.0484 |