Learn More
2',7'-Dichlorofluorescein diacetate, TRC
Product Code. 30134089
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134089 1 g 1g
30136155 50 mg 50mg
30009769 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134089 Supplier Toronto Research Chemicals Supplier No. TRCD4345331G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2044-85-1
Molecular Formula C24H14Cl2O7
Molecular Weight (g/mol) 485.27
Synonym MFCD 37501, 3',6'-Bis(acetyloxy)-2',7'-dichlorospiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one
SMILES CC(=O)Oc1cc2Oc3cc(OC(=O)C)c(Cl)cc3C4(OC(=O)c5ccccc45)c2cc1Cl
Specifications
Quantity 1 g
InChI Formula InChI=1S/C24H14Cl2O7/c1-11(27)30-21-9-19-15(7-17(21)25)24(14-6-4-3-5-13(14)23(29)33-24)16-8-18(26)22(31-12(2)28)10-20(16)32-19/h3-10H,1-2H3
Formula Weight 484.0117
Chemical Name or Material 2',7'-Dichlorofluorescein diacetate