missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethylcyclohexanone, cis + trans, 99%

Product Code. 11490493
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490493 25 g 25g
11400503 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490493 Supplier Thermo Scientific Alfa Aesar Supplier No. A12647.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2816-57-1 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2816-57-1
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00001637
InChI Key AILVYPLQKCQNJC-UHFFFAOYSA-N
Synonym 2,6-dimethylcyclohexanone, cyclohexanone, 2,6-dimethyl, cyclohexanone,6-dimethyl, 2,6-dimethyl cyclohexanone, 2,6-dimethyl-cyclohexanone, cyclohexanone,2,6-dimethyl, 2,6-dimethylcyclohexanone,c&t, 2,6-dimethyl-cyclohexan-1-one, 2,6-dimethylcyclohexanone cis + trans, 2,6-dimethylcyclohexanone, cis + trans
PubChem CID 17780
IUPAC Name 2,6-dimethylcyclohexan-1-one
SMILES CC1CCCC(C1=O)C

Specifications

Density 0.918
Boiling Point 174°C to 176°C
Flash Point 51°C (123°F)
Refractive Index 1.447
Quantity 25 g
UN Number UN1224
Beilstein 1099000
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 2,6-Dimethylcyclohexanone, cis + trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.