Learn More
2,6-Diiodo-4-nitrophenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Diiodo-4-nitrophenol |
| CAS | 305-85-1 |
| Molecular Formula | C6H3I2NO3 |
| Quantity | 500 mg |
| Synonym | 4-Nitro-2,6-diiodophenol, Ancylol, DNP, Disofen, Disophenol, Iodofen, Iodophen, Syngamix |
| InChI Formula | InChI=1S/C6H3I2NO3/c7-4-1-3(9(11)12)2-5(8)6(4)10/h1-2,10H |
| SMILES | O=[N](C1=CC(I)=C(O)C(I)=C1)=O |
| Molecular Weight (g/mol) | 390.9 |
| Formula Weight | 390.8202 |