missing translation for 'onlineSavingsMsg'
Learn More

2,6-Difluorobenzenesulfonyl chloride, 97%

Product Code. 11400144
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11400144 5 g 5g
11490134 1 g 1g
11410144 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11400144 Supplier Thermo Scientific Alfa Aesar Supplier No. A13595.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 60230-36-6 | C6H3ClF2O2S | 212.595 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60230-36-6
Molecular Formula C6H3ClF2O2S
Molecular Weight (g/mol) 212.595
MDL Number MFCD00085004
InChI Key QXWAUQMMMIMLTO-UHFFFAOYSA-N
Synonym 2,6-difluorobenzene-1-sulfonyl chloride, 2,6-difluorobenzenesulfonylchloride, 2,6-difluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 2,6-difluoro, 2,6-difluoro-benzenesulfonyl chloride, 2,6-difluorobenzene sulfonyl chloride, 2,6-difluorophenyl chlorosulfone, pubchem1957, acmc-209mil, ksc352q0t
PubChem CID 2734271
IUPAC Name 2,6-difluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)F)S(=O)(=O)Cl)F

Specifications

Density 1.568
Boiling Point 127°C to 130°C (6.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.526
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 212.6
Percent Purity 97%
Chemical Name or Material 2,6-Difluorobenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.