Learn More
2,6-Difluoro-4-methoxybenzoic acid, 97%, Thermo Scientific™
Product Code. 11382529
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 11382529
Brand: Thermo Scientific Alfa Aesar H26640.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 123843-65-2
Molecular Formula C8H6F2O3
Molecular Weight (g/mol) 188.13
MDL Number MFCD02949623
InChI Key LGHVYSAINQRZJQ-UHFFFAOYSA-N
Synonym benzoic acid, 2,6-difluoro-4-methoxy, 2,6-difluoro-4-methoxy-benzoic acid, acmc-1c2va, 2,6-difluoro-p-anisic acid, ksc495m7l, 4-carboxy-3,5-difluoroanisole, benzoic acid,2,6-difluoro-4-methoxy, 2,6-difluoro-4-methoxybenzoic acid
PubChem CID 2778228
IUPAC Name 2,6-difluoro-4-methoxybenzoic acid
SMILES COC1=CC(F)=C(C(O)=O)C(F)=C1
Specifications
Melting Point 183°C to 185°C
Quantity 1 g
Formula Weight 188.13
Percent Purity 97%
Chemical Name or Material 2,6-Difluoro-4-methoxybenzoic acid