missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dichlorobenzyl Alcohol, TRC

Product Code. 30179633
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179633 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30179633 Supplier Toronto Research Chemicals Supplier No. TRCD4322055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 15258-73-8
Quantity 5 g
Molecular Formula C7 H6 Cl2 O
Synonym (2,6-Dichlorophenyl)methanol, Benzenemethanol, 2,6-dichloro-, Benzyl alcohol, 2,6-dichloro- (8CI), 2,6-Dichlorobenzenemethanol, 2,6-Dichlorobenzyl alcohol, NSC 215733, 2,4-Dichlorobenzyl Alcohol Imp. B (EP)
InChI Formula InChI=1S/C7H6Cl2O/c8-6-2-1-3-7(9)5(6)4-10/h1-3,10H,4H2
SMILES OCc1c(Cl)cccc1Cl
IUPAC Name (2,6-dichlorophenyl)methanol
Molecular Weight (g/mol) 177.028
Formula Weight 175.9796
Chemical Name or Material 2,6-Dichlorobenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.