Learn More
2,6-Dichlorobenzamide, TRC
Product Code. 30386156
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386156 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30386156 Supplier Toronto Research Chemicals Supplier No. TRCD4317451G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,6-Dichlorobenzamide
CAS 2008-58-4
Molecular Formula C7 H5 Cl2 N O
Quantity 1 g
Synonym 2,6-Dichlorobenzamide, 2,6-BAM, BAM, NSC 53137, M-01
InChI Formula InChI=1S/C7H5Cl2NO/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3H,(H2,10,11)
SMILES NC(=O)c1c(Cl)cccc1Cl
IUPAC Name 2,6-dichlorobenzamide
Molecular Weight (g/mol) 190.03
Formula Weight 188.9748