Learn More
2,6-Dichloro-4-nitropyridine-1-oxide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,6-Dichloro-4-nitropyridine-1-oxide |
| CAS | 2587-01-1 |
| Molecular Formula | C5 H2 Cl2 N2 O3 |
| Quantity | 5 g |
| Synonym | Pyridine, 2,6-dichloro-4-nitro-, 1-oxide, 2,6-Dichloro-4-nitropyridine N-oxide, NSC 136570, 2,6-Dichloro-4-nitropyridine-1-oxide |
| InChI Formula | InChI=1S/C5H2Cl2N2O3/c6-4-1-3(9(11)12)2-5(7)8(4)10/h1-2H |
| SMILES | [O-][N+](=O)c1cc(Cl)[n+]([O-])c(Cl)c1 |
| IUPAC Name | 2,6-dichloro-4-nitro-1-oxidopyridin-1-ium |
| Molecular Weight (g/mol) | 208.99 |
| Formula Weight | 207.9442 |