missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-cyanophenyl Heptanoate-d13, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Bromoxynil Heptanoate-D13 |
| Molecular Formula | C14 D13 H2 Br2 N O2 |
| Quantity | 0.01 g |
| Synonym | 2,6-Dibromo-4-cyanophenyl heptanoate-d13, 2,6-Dibromo-4-cyanophenyl heptanoate-2,2,3,3,4,4,5,5,6,6,7,7,7-d13, Bromoxynil Heptanoate-D13 |
| InChI Formula | InChI=1 S/C14H15Br2NO2/c1-2-3-4-5-6-13(18)19-14-11(15)7-10(9-17)8-12(14)16/h7-8 H,2-6H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C(=O)Oc1c(Br)cc(cc1Br)C#N |
| IUPAC Name | (2,6-dibromo-4-cyanophenyl) 2,2,3,3,4,4,5,5,6,6,7,7,7-tridecadeuterioheptanoate |
| Molecular Weight (g/mol) | 402.1625 |
| Formula Weight | 400.0286 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction