missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-p-benzoquinone, TRC

Product Code. 30006743
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006743 25 g 25g
30006878 10 g 10g
30003493 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006743 Supplier Toronto Research Chemicals Supplier No. TRCD42943525G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 719-22-2
Molecular Formula C14 H20 O2
Molecular Weight (g/mol) 220.31
Synonym 2,6-Bis(1,1-dimethylethyl)-2,5-cyclohexadiene-1,4-dione, 2,6-Bis[1,1-dimethyl ethyl]quinone, 2,6-Di-tert-butyl-1,4-benzoquinone, 2,6-Di-tert-butyl-2,5-cyclohexadien-1,4-dione, 2,6-Di-tert-butyl-2,5-cyclohexadiene-1,4-dione, 2,6-Di-tert-butyl-p-benzoquinone, 2,6-Di-tert-butyl-p-quinone, 2,6-Di-tert-butylbenzoquinone, 2,6-Di-tert-butylquinone, 3,5-Di-tert-butyl-p-benzoquinone, 3,5-Di-tert-butylquinone, NSC 14448
IUPAC Name 2,6-ditert-butylcyclohexa-2,5-diene-1,4-dione
SMILES CC(C)(C)C1=CC(=O)C=C(C1=O)C(C)(C)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C14H20O2/c1-13(2,3)10-7-9(15)8-11(12(10)16)14(4,5)6/h7-8H,1-6H3
Formula Weight 220.1463
Chemical Name or Material 2,6-Di-tert-butyl-p-benzoquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.