Learn More
2-[(6-Chloro-4-pyrimidinyl)oxy]-Alpha-(dimethoxymethyl)benzeneacetic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-[(6-Chloro-4-pyrimidinyl)oxy]-alpha-(dimethoxymethyl)benzeneacetic Acid Methyl Ester |
| CAS | 143230-42-6 |
| Molecular Formula | C16H17ClN2O5 |
| Quantity | 5 mg |
| Synonym | Methyl 2-[2-(6-Chloropyrimidin-4-yloxy)phenyl]-3,3-dimethoxypropanoate |
| InChI Formula | InChI=1S/C16H17ClN2O5/c1-21-15(20)14(16(22-2)23-3)10-6-4-5-7-11(10)24-13-8-12(17)18-9-19-13/h4-9,14,16H,1-3H3 |
| SMILES | COC(OC)C(C(=O)OC)c1ccccc1Oc2cc(Cl)ncn2 |
| IUPAC Name | methyl 2-[2-(6-chloropyrimidin-4-yl)oxyphenyl]-3,3-dimethoxypropanoate |
| Molecular Weight (g/mol) | 352.77 |
| Formula Weight | 352.0826 |