Learn More
p-Tolyl Sulfate-d7 Potassium Salt, TRC
Click to view available options
Quantity:
1 mg
2.5 mg
25 mg
Unit Size:
1mg
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | p-Tolyl Sulfate-d7 Potassium Salt |
| Purity Grade Notes | HPLC |
| Molecular Formula | C7D7KO4S |
| Quantity | 2.5 mg |
| Synonym | 4-Cresol Sulfate-d7, p-Cresol Sulfate-d7 Potassium Salt, Mono(4-methylphenyl) Ester Sulfuric Acid-d7 Potassium Salt, Mono(p-tolyl) Ester Sulfuric Acid-d7 Potassium Salt |
| InChI Formula | InChI=1S/C7H8O4S.K/c1-6-2-4-7(5-3-6)11-12(8,9)10;/h2-5H,1H3,(H,8,9,10);/q;+1/p-1/i1D3,2D,3D,4D,5D; |
| SMILES | [K+].[2H]c1c([2H])c(c([2H])c([2H])c1OS(=O)(=O)[O-])C([2H])([2H])[2H] |
| IUPAC Name | potassium;[2,3,5,6-tetradeuterio-4-(trideuteriomethyl)phenyl] sulfate |
| Molecular Weight (g/mol) | 233.33 |
| Formula Weight | 233.0141 |
| Show More |