missing translation for 'onlineSavingsMsg'
Learn More

Calcitriol-d6, TRC

Product Code. 30333572
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 30333572

Brand: Toronto Research Chemicals TRCC1445022.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D6-1alpha,25-Dihydroxyvitamin D3 (Calcitriol-D6)
CAS 78782-99-7
Molecular Formula C27 2H6 H38 O3
Quantity 2.5 mg
Synonym 1,3-Cyclohexanediol, 4-methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R)-5-hydroxy-1-methyl-5-(methyl-d3)hexyl-6,6,6-d3]-7a-methyl-4H-inden-4-ylidene]ethylidene]-, (1R,3S,5Z)-, 9,10-Secocholesta-5,7,10(19)-triene-26,26,26,27,27,27-d6-1,3,25-triol, (1α,3β,5Z,7E)- (9CI), (1R,3S,5Z)-4-Methylene-5-[(2E)-2-[(1R,3aS,7aR)-octahydro-1-[(1R)-5-hydroxy-1-methyl-5-(methyl-d3)hexyl-6,6,6-d3]-7a-methyl-4H-inden-4-ylidene]ethylidene]-1,3-cyclohexanediol, 1α,25-Dihydroxyvitamin D3-[26,26,26,27,27,27]-2H6, Ro 21-5535/2, Calcitriol-d6, Calcitriol-26,26,26,27,27,27-D6, Vitamin D3-1alpha-25-dihydroxy D6 (26,26,26,27,27,27-D6)
InChI Formula InChI=1S/C27H44O3/c1-18(8-6-14-26(3,4)30)23-12-13-24-20(9-7-15-27(23,24)5)10-11-21-16-22(28)17-25(29)19(21)2/h10-11,18,22-25,28-30H,2,6-9,12-17H2,1,3-5H3/b20-10+,21-11-/t18-,22-,23-,24+,25+,27-/m1/s1/i3D3,4D3
SMILES [2H]C([2H])([2H])C(O)(CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C)C([2H])([2H])[2H]
IUPAC Name (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-7,7,7-trideuterio-6-hydroxy-6-(trideuteriomethyl)heptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol
Molecular Weight (g/mol) 422.67
Formula Weight 422.3667
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.