Learn More
2-iso-Propyl-d7-5-methyl-d3-phenol-3,4,6-d3, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Thymol-d13 |
| CAS | 1219798-93-2 |
| Molecular Formula | C10 2H13 H O |
| Quantity | 2.5 mg |
| Synonym | Thymol D13 (1-methylethyl D7) |
| InChI Formula | InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c(c(O)c([2H])c1C([2H])([2H])[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 2,4,5-trideuterio-6-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-3-(trideuteriomethyl)phenol |
| Molecular Weight (g/mol) | 163.3 |
| Formula Weight | 163.1861 |