Learn More
Fisher Chemical 2.5LT Hydrochloric acid, 36.5-38%, CertiFied ACS Plus
Brand:  Fisher Chemical A144-212
2.5LT Hydrochloric acid, 36.5-38%, CertiFied ACS Plus
Product Code. 16348247
  • ONLINE EXCLUSIVE PRICE
    112.00€
    108.66€ / 2.50 L
    Save 3.34€
    2% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Hydrochloric acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid