Learn More
N-Acetylneuraminic Acid Methyl Ester, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | N-Acetylneuraminic Acid Methyl Ester |
| CAS | 22900-11-4 |
| Molecular Formula | C12 H21 N O9 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C12H21NO9/c1-5(15)13-8-6(16)3-12(20,11(19)21-2)22-10(8)9(18)7(17)4-14/h6-10,14,16-18,20H,3-4H2,1-2H3,(H,13,15)/t6-,7+,8+,9+,10+,12-/m0/s1 |
| SMILES | COC(=O)[C@]1(O)C[C@H](O)[C@@H](NC(=O)C)[C@@H](O1)[C@H](O)[C@H](O)CO |
| IUPAC Name | methyl (2S,4S,5R,6R)-5-acetamido-2,4-dihydroxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylate |
| Molecular Weight (g/mol) | 323.3 |
| Formula Weight | 323.1216 |