missing translation for 'onlineSavingsMsg'
Learn More

Crospovidone (~40,000), TRC

Product Code. 30009621
Change view
Click to view available options
Quantity:
2.5 g
10 g
25 g
Unit Size:
10g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009621 2.5 g 2.5g
30136264 10 g 10g
30005238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009621 Supplier Toronto Research Chemicals Supplier No. TRCC8170002.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6 H9 N O)mon
Molecular Weight (g/mol) 111.14
Synonym α-Hydro-ω-hydropoly[1-(2-oxopyrrolidin-1-yl)ethylene], 2-Pyrrolidinone, 1-ethenyl-, homopolymer, 2-Pyrrolidinone, 1-vinyl-, polymers (8CI), 1-Vinyl-2-pyrrolidinone polymer, 1-Vinyl-2-pyrrolidone homopolymer, 1-Vinyl-2-pyrrolidone polymer, 1-Vinylpyrrolidin-2-one homopolymer, 143RP, 40K, 40K (vinyl polymer), 7TS13, ACP 10, AT 717, Agent AT 717, Agrimer 15, Agrimer 30, Agrimer 90, Agrimer ATF, Agrimer K 30, Albigen A, Aldacol Q, Antaron P 804, Antitox Vana, Aqua 65JL, Aqua-Quench EC/WE, Arostit TL 0280, B 7509, Bolinan, Cevian A 88036, Creejus K 30, Creejus K 50, Creejus K 90, Crospovidone, Cross P.V.P., Deltaspeed AF, Discol K 30L, Disintex 200, Divergan EF, Divergan F, Divergan RS, Epocros WS 560, Flexithix, Gaftex AE-K 15, Ganex P 804, HY 2, HY 3, Hemodesis, Hemodez, Hemodez N, Hemodez-N, Jarpol PVP-K 30, K 115, K 115 (vinyl polymer), K 12, K 120, K 15, K 15 (polymer), K 16-18, K 17, K 25, K 25 (surfactant), K 27/32, K 29-32, K 30, K 30C, K 30G, K 30PH, K 30W, K 40, K 50, K 50 (vinyl polymer), K 55, K 55 (vinyl polymer), K 60, K 60 (polymer), K 79, K 8, K 8 (vinyl polymer), K 85, K 85 (vinyl polymer), K 85N, K 85W, K 90, K 90HM, K 90W, K 92, K 92 (vinyl polymer), K88-96, Kollidon, Kollidon CE 50/50, Kollidon F 90, Kollidon K 12, Kollidon K 15, Kollidon K 17, Kollidon K 25, Kollidon K 29/32, Kollidon K 30, Kollidon K 60, Kollidon K 90, Kollidon K 90F, Kollidon R 90F, Konkrepol AD 1, Konkrepol B, LFC, Lumiten PPR 8450, Lumiten PR 8450, Luvicross M, Luviskol K, Luvitec K 15, Luvitec K 17, Luvitec K 30, Luvitec K 30PH, Luvitec K 40, Luvitec K 60, Luvitec K 80, Luvitec K 85, Luvitec K 90, Luvitec K 90PH, Luvitec PVP-K 30, MPK 90, MRTU 8457-79, N-Vinyl-2-pyrolidone homopolymer, N-Vinyl-2-pyrrolidinone homopolymer, N-Vinyl-2-pyrrolidone homopolymer, N-Vinyl-2-pyrrolidone polymer, N-Vinylbutyrolactam polymer, N-Vinylpyrrolidinone homopolymer, N-Vinylpyrrolidinone polymer, N-Vinylpyrrolidone homopolymer, N-Vinylpyrrolidone polymer, NKY-PVP-K, NP-K 30, NPK 15, NPK 90, Neocompensan, Neohemodes, P 0696, P-XL, PAK-K 15, PK 771, PV 03, PV 03 (vinyl pyrrolidone polymer), PV 05, PVK 30, PVP, PVP 10, PVP 1230, PVP 15, PVP 17PF, PVP 18, PVP 25, PVP 30, PVP 3000, PVP 360, PVP 360K, PVP 40, PVP 4000, PVP 40K, PVP 40T, PVP 50, PVP 99, PVP K 29/32, PVP-K, PVP-K 10, PVP-K 12, PVP-K 120, PVP-K 130, PVP-K 15, PVP-K 17, PVP-K 25, PVP-K 26/28, PVP-K 3, PVP-K 30, PVP-K 30W, PVP-K 35, PVP-K 40, PVP-K 45, PVP-K 60, PVP-K 64, PVP-K 70, PVP-K 78, PVP-K 79, PVP-K 80, PVP-K 85, PVP-K 88-96, PVP-K 90, PVP-K 90K95, PVP-K 95, PVPP, PVPP-XL 10, PX 4VP90, PX 90P, PX-K 30P, PX-K 90L, PX-K 90P, Peregal ST, Periston, Periston N, Peviston, Pitzcol K 120, Pitzcol K 17L, Pitzcol K 30, Pitzcol K 30L, Pitzcol K 50, Pitzcol K 90, Plasdone, Plasdone 4, Plasdone C, Plasdone XL, Plasdone XL 10, Plasmosan, Polidon, Poly(1-vinyl-2-pyrrolidinone), Poly(1-vinyl-2-pyrrolidone), Poly(1-vinylpyrrolidinone), Poly(N-vinyl-2-pyrrolidinone), Poly(N-vinyl-2-pyrrolidone), Poly(N-vinyl-2-pyrrolidone), Poly(N-vinyl-γ-butyrolactam), Poly(N-vinylbutyrolactam), Poly(N-vinylpyrrolidinone), Poly(N-vinylpyrrolidone), Poly(vinylpyrrolidinone), Poly(vinylpyrrolidone), Polyclar AT, Polyclar H, Polyclar L, Polyplasdone, Polyplasdone INF 10, Polyplasdone R-XL, Polyplasdone XL, Polyplasdone XL 10, Polyvidon, Polyvidon XL, Polyvidone, Polyvidone 25, Polyvidone K 30, Polyvinyl polypyrrolidone, Polyvinylpyrrolidon XL, Polyvinylpyrrolidone, Poviderm SK 3, Povidone, Povidone 30, Povidone K 12, Povidone K 15, Povidone K 17, Povidone K 25, Povidone K 29-32, Povidone K 30, Povidone K 90, Povidone K 90D, Protagent, Pvpon K 32, Rubiscole K 30, SD 13, SD 13 (polymer), Sokalan HP 165, Sokalan HP 50, Sokalan HP 53, Sokalan HP 60, Sokalan K 17P, Sokalan K 85P, Sokalan K 90, Sokalan K 90P, Sokolan HP 165, Sokolan HP 53, Sokolan HP 60, Sokolan K 30P, Sokolan K 85P, Subtosan, TSP 9A, Vinisil, Vinylpyrrolidinone polymer, Vinylpyrrolidone homopolymer, Vinylpyrrolidone polymer, ViviPrint 530, YK 001
IUPAC Name 1-vinylpyrrolidin-2-one
SMILES C=CN1CCCC1=O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2
Formula Weight 111.0684
Chemical Name or Material Povidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.