Learn More
2,5-Toluenediamine-d6 Dihydrochloride, TRC
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
Specifications
Specifications
| Chemical Name or Material | 2,5-Toluenediamine-d6 Dihydrochloride |
| Molecular Formula | C7H6D6Cl2N2 |
| Quantity | 50 mg |
| Synonym | Toluene-d6-2,5-diamine Dihydrochloride, 1,4-Diamino-2-methylbenzene-d6 Dihydrochloride, 1-Methyl-2,5-diaminobenzene-d6 Dihydrochloride, 2,5-Diaminotoluene-d6 Dihydrochloride, 2,5-Diaminotoluol-d6 Dihydrochloride, 2-Methyl-1,4-benzene-d6-diamine Dihydrochloride, 2-Methyl-1,4-phenyl-d6-enediamine Dihydrochloride, 2-Methyl-p-phenyl-d6-enediamine Dihydrochloride, 4-Amino-2-methylaniline-d6 Dihydrochloride, 4-Amino-3-methylaniline-d6 Dihydrochloride, C.I. 76042-d6 Dihydrochloride, Toluylene-d6-2,5-diamine Dihydrochloride |
| InChI Formula | InChI=1S/C7H10N2.2ClH/c1-5-4-6(8)2-3-7(5)9;;/h2-4H,8-9H2,1H3;2*1H/i1D3,2D,3D,4D;; |
| SMILES | NC1=C(C([2H])=C(C([2H])=C1[2H])N)C([2H])([2H])[2H].[2HCl] |
| IUPAC Name | 2-(methyl-d3)benzene-3,5,6-d3-1,4-diamine dihydrochloride |
| Molecular Weight (g/mol) | 201.13 |
| Formula Weight | 200.08 |