Learn More
2,5-Pyridinedicarboxylic Acid, TRC
Product Code. 30410346
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
missing translation for 'unitSize'
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30410346 25 g -
30413634 50 g 50g
30410855 10 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30410346 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCP99164025G
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 100-26-5
Molecular Formula C7 H5 N O4
Molecular Weight (g/mol) 167.12
Synonym Pyridine-2,5-dicarboxylic Acid, 2,5-Pyridinedicarboxylic acid, 2,5-Dicarboxypyridine, Isocinchomeronic acid, NSC 177
IUPAC Name pyridine-2,5-dicarboxylic acid
SMILES OC(=O)c1ccc(nc1)C(=O)O
Spezifikation
Quantity 25 g
InChI Formula InChI=1S/C7H5NO4/c9-6(10)4-1-2-5(7(11)12)8-3-4/h1-3H,(H,9,10)(H,11,12)
Formula Weight 167.0219
Chemical Name or Material Isocinchomeronic Acid