Learn More
2,5-Pyridinedicarboxylic Acid-13C7, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Pyridinedicarboxylic Acid-13C7 |
| CAS | 1189695-39-3 |
| Molecular Formula | 13C7H5NO4 |
| Quantity | 5 mg |
| Synonym | 2,5-Dicarboxypyridine-13C7, Isocinchomeronic Acid-13C7, NSC 177-13C7 |
| InChI Formula | InChI=1S/C7H5NO4/c9-6(10)4-1-2-5(7(11)12)8-3-4/h1-3H,(H,9,10)(H,11,12)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1 |
| SMILES | O[13C](=O)[13c]1[13cH][13cH][13c](n[13cH]1)[13C](=O)O |
| IUPAC Name | (2,3,4,5,6-^{13}C_{5})pyridine-2,5-dicarboxylic acid |
| Molecular Weight (g/mol) | 174.07 |
| Formula Weight | 174.0453 |