Learn More
2,5-Dioxopyrrolidine Sacubitril, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Dioxopyrrolidine Sacubitril |
| CAS | 1038924-97-8 |
| Molecular Formula | C24H27NO4 |
| Quantity | 25 mg |
| Synonym | (2R,4S)-5-(Biphenyl-4-yl)-4-(2,5-dioxopyrrolidin-1-yl)-2-methylpentanoic Acid Ethyl Ester, 2,5-Dioxopyrrolidine Sacubitril, Sacubitril Impurity 2 |
| InChI Formula | InChI=1S/C24H27NO4/c1-3-29-24(28)17(2)15-21(25-22(26)13-14-23(25)27)16-18-9-11-20(12-10-18)19-7-5-4-6-8-19/h4-12,17,21H,3,13-16H2,1-2H3/t17-,21+/m1/s1 |
| SMILES | CCOC(=O)[C@H](C)CC(Cc1ccc(cc1)c2ccccc2)N3C(=O)CCC3=O |
| Molecular Weight (g/mol) | 393.48 |
| Formula Weight | 393.194 |