Learn More
2,5-Dimethyl-2-hexanol, 98%
Product Code. 11414247
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414247 5 g 5g
11424247 25 g 25g
11434247 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11414247 Supplier Thermo Scientific Alfa Aesar Supplier No. B20352.06
Request Bulk or Custom Format
CAS: 3730-60-7 | C8H18O | 130.23 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3730-60-7
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00021814
InChI Key JPUIYNHIEXIFMV-UHFFFAOYSA-N
Synonym 2,5-dimethyl-2-hexanol, 2-hexanol, 2,5-dimethyl-, s, 2-hexanol, 2,5-dimethyl, 2-hexanol, 2,5-dimethyl-, s-+, acmc-209is7, 2-hexanol,2,5-dimethyl, 2-hexanol,5-dimethyl-, s, 2-hexanol,5-dimethyl-, s-+
PubChem CID 19506
IUPAC Name 2,5-dimethylhexan-2-ol
SMILES CC(C)CCC(C)(C)O
Specifications
Boiling Point 155°C to 156°C
Flash Point 52°C (125°F)
Refractive Index 1.422
Quantity 5 g
UN Number UN1987
Beilstein 1732761
Formula Weight 130.23
Percent Purity 98%
Chemical Name or Material 2,5-Dimethyl-2-hexanol