Learn More
2,5-Dihydroxybenzoic Acid-d3 (d2 Major), TRC
Product Code. 17977989
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17977989 5 mg 5mg
17917179 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17977989 Supplier Toronto Research Chemicals Supplier No. TRCD4516575MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 294661-02-2
Molecular Formula C7H3D3O4
Molecular Weight (g/mol) 157.14
Synonym 5-Hydroxysalicylic Acid-d3, Carboxyhydroquinone-d3, Gensigen-d3, Gensigon-d3, Gentisinic Acid-d3, Hydroquinonecarboxylic Acid-d3, NSC 27224-d3, NSC 49098-d3, NSC 78825-d3;
SMILES OC1=C(C(O)=O)C([2H])=C(O)C([2H])=C1[2H]
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C7H6O4/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,8-9H,(H,10,11)/i1D,2D,3D
Chemical Name or Material 2,5-Dihydroxybenzoic Acid-d3