missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS], TRC

Product Code. 30371544
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371544 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371544 Supplier Toronto Research Chemicals Supplier No. TRCD679195100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,5-Dihydroxybenzoic Acid Butylamine Salt [Matrix for MALDI-TOF/MS]
CAS 666174-80-7
Molecular Formula C7H6O4 . C4H11N
Quantity 100 mg
Synonym Benzoic acid, 2,5-dihydroxy-, compd. with 1-butanamine (1:1) (9CI, ACI), 2,5-Dihydroxybenzoic acid-butylamine (1:1) salt, Butylammonium 2,5-dihydroxybenzoate, 2,5-Dihydroxybenzoic acid butylamine salt, 2,5-Dihydroxybenzoic acid 1-butylamine salt
InChI Formula InChI=1S/C7H6O4.C4H11N/c8-4-1-2-6(9)5(3-4)7(10)11;1-2-3-4-5/h1-3,8-9H,(H,10,11);2-5H2,1H3
SMILES CCCCN.OC(=O)c1cc(O)ccc1O
IUPAC Name butan-1-amine;2,5-dihydroxybenzoic acid
Molecular Weight (g/mol) 227.26
Formula Weight 227.1158
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.