Learn More
2,5-Difluorophenyl isocyanate, 98%
Product Code. 11382837
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382837 250 mg 250mg
11372837 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11382837 Supplier Thermo Scientific Alfa Aesar Supplier No. L15996.MD
Request Bulk or Custom Format
CAS: 39718-32-6 | C7H3F2NO | 155.104 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 39718-32-6
Molecular Formula C7H3F2NO
Molecular Weight (g/mol) 155.104
MDL Number MFCD00013843
InChI Key SNHIIFOXCRYGGY-UHFFFAOYSA-N
PubChem CID 2733266
IUPAC Name 1,4-difluoro-2-isocyanatobenzene
SMILES C1=CC(=C(C=C1F)N=C=O)F
Specifications
Density 1.323
Boiling Point 78°C (33 mmHg)
Flash Point 56°C (132°F)
Refractive Index 1.494
Quantity 250 mg
UN Number UN3080
Beilstein 2091753
Sensitivity Moisture sensitive
Synonym 2,5-difluorophenyl isocyanate, 2,5-difluorophenylisocyanate, benzene, 1,4-difluoro-2-isocyanato, isocyanic acid 2,5-difluorophenyl ester, 2,5-difluorobenzenisocyanate, pubchem5022, acmc-1ahno, 2,5-difluoro-phenyl-isocyanate, benzene,1,4-difluoro-2-isocyanato
Formula Weight 155.11
Percent Purity 98%
Chemical Name or Material 2,5-Difluorophenyl isocyanate