missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Difluorobenzyl bromide, 98%
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Especificaciones
| Density | 1.609 |
| Refractive Index | 1.526 |
| Quantity | 50 g |
| UN Number | UN3265 |
| Beilstein | 7089248 |
| Formula Weight | 207.02 |
| Percent Purity | 98% |
| Chemical Name or Material | 2,5-Difluorobenzyl bromide |
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido