Learn More
2,5-Dichloroaniline D3 (ring D3), Dr. Ehrenstorfer
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | 2,5-Dichloroaniline-3,4,6-d3 |
| CAS | 783321-80-2 |
| Molecular Formula | C6H2D3Cl2N |
| Quantity | 10 mg |
| Synonym | 2,5-Dichloroaniline D3 (Ring D3), Benzen-2,4,5-d3-amine, 3,6-dichloro- (9CI) |
| InChI Formula | InChI=1S/C6H5Cl2N/c7-4-1-2-5(8)6(9)3-4/h1-3H,9H2/i1D,2D,3D |
| SMILES | [2H]c1c([2H])c(Cl)c(N)c([2H])c1Cl |
| IUPAC Name | 2,5-dichloro-3,4,6-trideuterioaniline |
| Molecular Weight (g/mol) | 165.04 |
| Formula Weight | 163.9987 g/mol |