Learn More
2,5-Anhydro-1,3-O-isopropylidene-6-O-trityl-D-glucitol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,5-Anhydro-1,3-O-isopropylidene-6-O-trityl-D-glucitol |
| CAS | 65758-50-1 |
| Molecular Formula | C27 H28 O5 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C27H28O5/c1-26(2)31-24-23(28)22(30-25(24)32-26)18-29-27(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-17,22-25,28H,18H2,1-2H3/t22-,23+,24-,25-/m0/s1 |
| SMILES | CC1(C)O[C@@H]2O[C@@H](COC(c3ccccc3)(c4ccccc4)c5ccccc5)[C@@H](O)[C@@H]2O1 |
| IUPAC Name | (3aS,5S,6R,6aS)-2,2-dimethyl-5-(trityloxymethyl)-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]dioxol-6-ol |
| Molecular Weight (g/mol) | 432.51 |
| Formula Weight | 432.1937 |