Learn More
2,4-Octadienal, predominantly trans,trans (Technical Grade), TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Octadienal','predominantly trans,trans (Technical Grade) |
| CAS | 30361-28-5 |
| Molecular Formula | C8H12O |
| Quantity | 500 mg |
| Synonym | (2E,4E)-2,4-Octadienal, (2E,4E)-Octa-2,4-dienal, (2E,4E)-Octadienal, (E,E)-2,4-Octadien-1-al, (E,E)-2,4-Octadienal, 2-trans-4-trans-Octadienal, E,E-2,4-Octadienal, all-trans-2,4-Octadienal, trans,trans-Octa-2,4-dienal, trans-2,trans-4-Octadienal |
| InChI Formula | InChI=1S/C8H12O/c1-2-3-4-5-6-7-8-9/h4-8H,2-3H2,1H3/b5-4+,7-6+ |
| SMILES | CCC\C=C\C=C\C=O |
| IUPAC Name | (2E,4E)-octa-2,4-dienal |
| Molecular Weight (g/mol) | 124.18 |
| Formula Weight | 124.0888 |