Learn More
2-(4-Nitro-1-oxoisoindolin-2-yl)pentanedioic Acid, TRC
Product Code. 30471463
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30471463 100 mg 100mg
30469004 50 mg -
30466423 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30471463 Supplier Toronto Research Chemicals Supplier No. TRCN496903100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 295357-72-1
Molecular Formula C13H12N2O7
Molecular Weight (g/mol) 308.25
Synonym 2-(1,3-Dihydro-4-nitro-1-oxo-2H-isoindol-2-yl)pentanedioic Acid
IUPAC Name 2-(7-nitro-3-oxo-1H-isoindol-2-yl)pentanedioic acid
SMILES OC(=O)CCC(N1Cc2c(cccc2[N+](=O)[O-])C1=O)C(=O)O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C13H12N2O7/c16-11(17)5-4-10(13(19)20)14-6-8-7(12(14)18)2-1-3-9(8)15(21)22/h1-3,10H,4-6H2,(H,16,17)(H,19,20)
Formula Weight 308.0645
Chemical Name or Material 2-(4-Nitro-1-oxoisoindolin-2-yl)pentanedioic Acid