missing translation for 'onlineSavingsMsg'
Learn More

2'-(4-Methylumbelliferyl)-Alpha-D-N-acetylneuraminic Acid Sodium Salt, TRC

Product Code. 30012057
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012057 25 mg 25mg
30011834 10 mg 10mg
30008954 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012057 Supplier Toronto Research Chemicals Supplier No. TRCM33420025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76204-02-9
Molecular Formula C21 H24 N O11 . Na
Molecular Weight (g/mol) 489.41
Synonym α-Neuraminic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl 5-(acetylamino)-3,5-dideoxy-, sodium salt (1:1), D-glycero-α-D-galacto-2-Nonulopyranosidonic acid, 4-methyl-2-oxo-2H-1-benzopyran-7-yl 5-(acetylamino)-3,5-dideoxy-, monosodium salt, α-Neuraminic acid, N-acetyl-2-O-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-, monosodium salt (9CI), 2-(4-Methylumbelliferyl)-N-acetylneuraminic acid sodium salt, 4-MUNANA, MUNANA, Sodium 2-(4-methylumbelliferyl)-N-acetylneuraminate, 2'-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt, N-Acetyl-2-O-(4-methyl-2-oxo-2H-1-benzopyran-7-yl)-α-neuraminic acid sodium salt, 2-O-4-Methylumbelliferyl-N-acetyl-α-D-neuraminic acid sodium salt, 4-Methylumbelliferyl α-NeuAc sodium, 4-Methylumbelliferyl-N-acetyl-α-D-neuraminic acid sodium salt, N-Acetyl-2-O-(4-methylumbelliferyl)-α-neuraminic acid sodium salt
IUPAC Name sodium;(2R,4S,5R,6R)-5-acetamido-4-hydroxy-2-(4-methyl-2-oxochromen-7-yl)oxy-6-[(1R,2R)-1,2,3-trihydroxypropyl]oxane-2-carboxylate
SMILES [Na+].CC(=O)N[C@@H]1[C@@H](O)C[C@](Oc2ccc3C(=CC(=O)Oc3c2)C)(O[C@H]1[C@H](O)[C@H](O)CO)C(=O)[O-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C21H25NO11.Na/c1-9-5-16(27)31-15-6-11(3-4-12(9)15)32-21(20(29)30)7-13(25)17(22-10(2)24)19(33-21)18(28)14(26)8-23;/h3-6,13-14,17-19,23,25-26,28H,7-8H2,1-2H3,(H,22,24)(H,29,30);/q;+1/p-1/t13-,14+,17+,18+,19+,21-;/m0./s1
Formula Weight 489.1247
Percent Purity >95%
Chemical Name or Material 2'-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.