missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methylpiperazino)benzaldehyde, ≥95%, Thermo Scientific™

Product Code. 10104154
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10104154

Brand: Thermo Scientific Maybridge CC36004EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 85803-62-9
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD01313808
InChI Key GSRYZPWIWYYROI-UHFFFAOYSA-N
Synonym 2-4-methylpiperazin-1-yl benzaldehyde, 2-4-methylpiperazino benzaldehyde, benzaldehyde,2-4-methyl-1-piperazinyl, 4-methylpiperazin-1-ylbenzaldehyde, 2-4-methylpiperazinyl benzaldehyde, 2-4-methylpiperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl benzaldehyde, 2-4methyl-1-piperazinyl-benzaldehyde, 2-4-methyl-1-piperazinyl-benzaldehyde, 2-4-methylpiperazin-1-yl-benzaldehyde
PubChem CID 2795566
IUPAC Name 2-(4-methylpiperazin-1-yl)benzaldehyde
SMILES CN1CCN(CC1)C2=CC=CC=C2C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 10 g
Formula Weight 204.27
Percent Purity 95%
Chemical Name or Material 2-(4-Methylpiperazin-1-yl)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.