Learn More
2-(4-Methylphenyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole, 97%, Thermo Scientific™
Product Code. 15484526
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484526 10 g 10g
15474526 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15484526 Supplier Thermo Scientific Alfa Aesar Supplier No. H58786.09
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1421262-98-7
Molecular Formula C20H19NS
Molecular Weight (g/mol) 305.439
MDL Number MFCD22683014
InChI Key SZRKBHGWZOAMML-UHFFFAOYSA-N
Synonym 2-4-methylphenyl-4-5,6,7,8-tetrahydronaphthalen-2-yl-1,3-thiazole, 2-4-methylphenyl-4-5,6,7,8-tetrahydro-2-naphthyl thiazole
PubChem CID 97030363
IUPAC Name 2-(4-methylphenyl)-4-(5,6,7,8-tetrahydronaphthalen-2-yl)-1,3-thiazole
SMILES CC1=CC=C(C=C1)C2=NC(=CS2)C3=CC4=C(CCCC4)C=C3
Specifications
Quantity 10 g
Formula Weight 305.44
Percent Purity 97%
Chemical Name or Material 2-(4-Methylphenyl)-4-(5,6,7,8-tetrahydro-2-naphthyl)thiazole