missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Methoxyphenyl)ethylamine Hydrochloride, TRC

Product Code. 30471751
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30471751 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30471751 Supplier Toronto Research Chemicals Supplier No. TRCM266115100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(4-Methoxyphenyl)ethylamine Hydrochloride
CAS 645-58-9
Molecular Formula C9H14ClNO
Quantity 100 g
Synonym 4-Methoxybenzeneethanamine Hydrochloride, p-Methoxyphenethylamine Hydrochloride, 2-(4-Methoxyphenyl)ethanamine Hydrochloride, 2-(4-Methoxyphenyl)ethylamine Hydrochloride, 2-(p-Methoxyphenyl)ethylamine Hydrochloride, 4-Methoxy-beta-phenethylamine Hydrochloride, 4-Methoxy-beta-phenylethylamine Hydrochloride, 4-Methoxybenzeneethanamine Hydrochloride, 4-Methoxyphenethylamine Hydrochloride, 4-Methoxyphenylethylamine Hydrochloride, Homoanisylamine, NSC 43687 Hydrochloride, O-Methyltyramine Hydrochloride, Tyramine Methyl Ether Hydrochloride, O-Methyltyramine Hydrochloride, [2-[4-(Methyloxy)phenyl]ethyl]amine Hydrochloride, p-Methoxyphenethylamine Hydrochloride
InChI Formula InChI=1S/C9H13NO.ClH/c1-11-9-4-2-8(3-5-9)6-7-10;/h2-5H,6-7,10H2,1H3;1H
SMILES Cl.COc1ccc(CCN)cc1
IUPAC Name 2-(4-methoxyphenyl)ethanamine;hydrochloride
Molecular Weight (g/mol) 187.67
Formula Weight 187.0764
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.