missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Methoxyphenyl)ethylamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(4-Methoxyphenyl)ethylamine Hydrochloride |
| CAS | 645-58-9 |
| Molecular Formula | C9H14ClNO |
| Quantity | 250 g |
| Synonym | 4-Methoxybenzeneethanamine Hydrochloride, p-Methoxyphenethylamine Hydrochloride, 2-(4-Methoxyphenyl)ethanamine Hydrochloride, 2-(4-Methoxyphenyl)ethylamine Hydrochloride, 2-(p-Methoxyphenyl)ethylamine Hydrochloride, 4-Methoxy-beta-phenethylamine Hydrochloride, 4-Methoxy-beta-phenylethylamine Hydrochloride, 4-Methoxybenzeneethanamine Hydrochloride, 4-Methoxyphenethylamine Hydrochloride, 4-Methoxyphenylethylamine Hydrochloride, Homoanisylamine, NSC 43687 Hydrochloride, O-Methyltyramine Hydrochloride, Tyramine Methyl Ether Hydrochloride, O-Methyltyramine Hydrochloride, [2-[4-(Methyloxy)phenyl]ethyl]amine Hydrochloride, p-Methoxyphenethylamine Hydrochloride |
| InChI Formula | InChI=1S/C9H13NO.ClH/c1-11-9-4-2-8(3-5-9)6-7-10;/h2-5H,6-7,10H2,1H3;1H |
| SMILES | Cl.COc1ccc(CCN)cc1 |
| IUPAC Name | 2-(4-methoxyphenyl)ethanamine;hydrochloride |
| Molecular Weight (g/mol) | 187.67 |
| Formula Weight | 187.0764 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?