Learn More
2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™
Product Code. 15410686
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410686 250 mg 250mg
15490676 1 g 1g
15400686 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15410686 Supplier Thermo Scientific Alfa Aesar Supplier No. H54038.MD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 54001-16-0
Molecular Formula C12H11NO3S
Molecular Weight (g/mol) 249.284
MDL Number MFCD06857928
InChI Key HWHHZTHVYFDJNK-UHFFFAOYSA-N
Synonym 2-4-methoxyphenyl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-4-methoxyphenyl-4-methylthiazole-5-carboxylic acid, 2-4-methoxyphenyl-4-methyl-5-thiazolecarboxylic acid, 4-methyl-2-4-methoxyphenyl thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2-4-methoxyphenyl-4-methyl, 2-4-methoxy phenyl-4-methyl-1,3-thiazol-5-carboxylic acid
PubChem CID 4962843
IUPAC Name 2-(4-methoxyphenyl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC=C(C=C2)OC)C(=O)O
Specifications
Melting Point 236°C to 241°C
Quantity 250 mg
Formula Weight 249.28
Percent Purity 97%
Chemical Name or Material 2-(4-Methoxyphenyl)-4-methylthiazole-5-carboxylic acid