Learn More
2-(4-methoxyphenoxy)ethanamine hydrochloride, 97%, Thermo Scientific™
Product Code. 10230323
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 10230323
Brand: Thermo Scientific Maybridge BTB10107EA
Request Bulk or Custom Format
Chemical Identifiers
CAS 98959-77-4
Molecular Formula C9H14ClNO2
Molecular Weight (g/mol) 203.666
InChI Key YCFXTJDWVNEVEX-UHFFFAOYSA-N
Synonym 2-4-methoxyphenoxy ethan-1-amine hydrochloride, 2-4-methoxyphenoxy ethanamine hydrochloride, 2-4-methoxyphenoxy ethylamine hydrochloride, 2-4-methoxyphenoxy ethylaminehydrochloride, 1-2-aminoethoxy-4-methoxybenzene hydrochloride
PubChem CID 43810691
IUPAC Name 2-(4-methoxyphenoxy)ethanamine;hydrochloride
SMILES COC1=CC=C(C=C1)OCCN.Cl
Specifications
Assay Percent Range 97%
Quantity 10 g
Formula Weight 203.67
Chemical Name or Material 2-(4-methoxyphenoxy)ethanamine hydrochloride