Learn More
2,4-Dinitrobenzyl Bromide, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dinitrobenzyl Bromide |
| CAS | 3013-38-5 |
| Molecular Formula | C7 H5 Br N2 O4 |
| Quantity | 1 g |
| Synonym | 1-(Bromomethyl)-2,4-dinitrobenzene (ACI), Toluene, α-bromo-2,4-dinitro- (6CI, 7CI, 8CI), 2,4-Dinitrobenzyl bromide, o,p-Dinitrobenzyl bromide |
| InChI Formula | InChI=1S/C7H5BrN2O4/c8-4-5-1-2-6(9(11)12)3-7(5)10(13)14/h1-3H,4H2 |
| SMILES | [O-][N+](=O)c1ccc(CBr)c(c1)[N+](=O)[O-] |
| IUPAC Name | 1-(bromomethyl)-2,4-dinitrobenzene |
| Molecular Weight (g/mol) | 261.03 |
| Formula Weight | 259.943 |