Learn More
2,4-Dimethylphenoxyacetic acid, 98+%, Thermo Scientific™
Product Code. 11324544
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 11324544
Brand: Thermo Scientific Alfa Aesar L01296.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13334-49-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00014355
InChI Key XRTZHWXWLUGOAT-UHFFFAOYSA-N
Synonym 2-2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxy acetic acid, 2,4-dimethylphenoxyacetic acid, 2,4-xylyloxyacetic acid, 2-2,4-dimethylphenoxy ethanoic acid, 2,4-dimethylphenoxyaceticacid, akos bbb/200, akos bc-2696, acmc-1btqb, labotest-bb lt00847868
PubChem CID 83354
IUPAC Name 2-(2,4-dimethylphenoxy)acetic acid
SMILES CC1=CC(=C(C=C1)OCC(=O)O)C
Specifications
Melting Point 139°C to 143°C
Quantity 5 g
Beilstein 2263832
Formula Weight 180.2
Percent Purity ≥98%
Chemical Name or Material 2,4-Dimethylphenoxyacetic acid