missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethylphenol-3,5,6-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dimethylphenol-3,5,6-D3 |
| CAS | 93951-75-8 |
| Molecular Formula | C8 2H3 H7 O |
| Quantity | 0.25 g |
| Synonym | Phen-2,3,5-d3-ol, 4,6-dimethyl- (9 CI), 2,4-Dimethyl-3,5,6-trideuteriophenol, 2,4-Dimethylphenol-3,5,6-D3, 2,4-Dimethylphenol-3,5,6-d3, 2,4-Dimethylphenol (ring-D3) |
| InChI Formula | InChI=1 S/C8H10O/c1-6-3-4-8(9)7(2)5-6/h3-5,9 H,1-2H3/i3D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c(O)c(C)c([2 H])c1C |
| IUPAC Name | 2,3,5-trideuterio-4,6-dimethylphenol |
| Molecular Weight (g/mol) | 125.18 |
| Formula Weight | 125.092 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction