Learn More
2,4-Dimethylbenzoyl chloride, 97%, Thermo Scientific™
Product Code. 11315379
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315379 1 g 1g
11325379 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11315379 Supplier Thermo Scientific Alfa Aesar Supplier No. H50387.03
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21900-42-5
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD00798005
InChI Key JUOGRBULLOEKKQ-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,4-dimethyl, 2,4-dimethyl-benzoyl chloride, benzoyl chloride,2,4-dimethyl, acmc-20aocb, 2.4-dimethylbenzoylchlorid, 2,4-di-methylbenzoyl chloride, 2,4-dimethyl benzoyl chloride, buttpark 48\06-96, 2,4-dimethyl-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethyl-6ci,8ci,9ci
PubChem CID 140867
IUPAC Name 2,4-dimethylbenzoyl chloride
SMILES CC1=CC(=C(C=C1)C(=O)Cl)C
Specifications
Melting Point 24°C to 26°C
Quantity 1 g
UN Number UN3265
Boiling Point 82°C (1 mmHg)
Formula Weight 168.62
Percent Purity 97%
Chemical Name or Material 2,4-Dimethylbenzoyl chloride