missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dihydroxybenzophenone-2',3',4',5',6'-d5, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Specifications
Specifications
| Chemical Name or Material | 2,4-Dihydroxybenzophenone-d5 |
| CAS | 91586-06-0 |
| Molecular Formula | C13 D5 H5 O3 |
| Quantity | 0.05 g |
| Synonym | 2,4-Dihydroxybenzophenone D5 (Phenyl D5) |
| InChI Formula | InChI=1 S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15 H/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)c2ccc(O)cc2O)c([2 H])c1[2 H] |
| IUPAC Name | (2,4-dihydroxyphenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanone |
| Molecular Weight (g/mol) | 219.248 |
| Formula Weight | 219.094 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction