Learn More
(2,4-Dichlorophenoxy-d3)acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-D D3 |
| CAS | 202480-67-9 |
| Molecular Formula | C8 2H3 H3 Cl2 O3 |
| Quantity | 5 mg |
| Synonym | Acetic acid, (4,6-dichlorophenoxy-2,3,5-d3)- (9CI), 2,4-D D3, (2,4-Dichloro-3,5,6-trideuteriophenoxy)acetic Acid |
| InChI Formula | InChI=1S/C8H6Cl2O3/c9-5-1-2-7(6(10)3-5)13-4-8(11)12/h1-3H,4H2,(H,11,12)/i1D,2D,3D |
| SMILES | [2H]c1c([2H])c(OCC(=O)O)c(Cl)c([2H])c1Cl |
| IUPAC Name | 2-(2,4-dichloro-3,5,6-trideuteriophenoxy)acetic acid |
| Molecular Weight (g/mol) | 224.06 |
| Formula Weight | 222.9882 |