Learn More
2,4-Dichloro-6-isopropylamino-1,3,5-triazine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-6-isopropylamino-1,3,5-triazine |
| CAS | 3703-10-4 |
| Molecular Formula | C7 H9 F N2 O2 S |
| Quantity | 500 mg |
| Synonym | 4,6-Dichloro-N-(1-methylethyl)-1,3,5-triazin-2-amine, 2,4-Dichloro-6-(isopropylamino)-s-triazine, 2,4-Dichloro-6-[(1-methylethyl)amino]-1,3,5-triazine, 2,4-Dichloro-6-isopropylamino-s-triazine, 2,6-Dichloro-4-isopropylamino-s-triazine, N-Isopropylaminodichlorotriazine, NSC 344238, s-Dichloroisopropylaminotriazine |
| InChI Formula | InChI=1S/C7H9FN2O2S/c1-4-2-7(13(10,11)12)5(8)3-6(4)9/h2-3H,9H2,1H3,(H2,10,11,12) |
| SMILES | Cc1cc(c(F)cc1N)S(=O)(=O)N |
| IUPAC Name | 4-amino-2-fluoro-5-methylbenzenesulfonamide |
| Molecular Weight (g/mol) | 204.222 |
| Formula Weight | 204.037 |