Learn More
2,4-Dichloro-5-trifluoromethylpyrimidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-5-trifluoromethylpyrimidine |
| CAS | 3932-97-6 |
| Molecular Formula | C5HCl2F3N2 |
| Quantity | 5 g |
| Synonym | 2,6-Dichloro-5-trifluoromethylpyrimidine, 5-Trifluoromethyl-2,4-dichloropyrimidine |
| InChI Formula | InChI=1S/C5HCl2F3N2/c6-3-2(5(8,9)10)1-11-4(7)12-3/h1H |
| SMILES | ClC1=NC(Cl)=C(C(F)(F)F)C=N1 |
| IUPAC Name | 2,4-dichloro-5-(trifluoromethyl)pyrimidine |
| Molecular Weight (g/mol) | 216.98 |
| Formula Weight | 215.95 |