Learn More
2,4-Diacetylphloroglucinol, TRC
Specifications
Specifications
| Chemical Name or Material | 2,4-Diacetylphloroglucinol |
| CAS | 2161-86-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 H10 O5 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C10H10O5/c1-4(11)8-6(13)3-7(14)9(5(2)12)10(8)15/h3,13-15H,1-2H3 |
| SMILES | CC(=O)c1c(O)cc(O)c(C(=O)C)c1O |
| IUPAC Name | 1-(3-acetyl-2,4,6-trihydroxyphenyl)ethanone |
| Molecular Weight (g/mol) | 210.18 |
| Formula Weight | 210.0528 |
| Show More |